Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Acetyl-CoA Carboxylase (Ser80) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538107
Description
Catalyzes the rate-limiting reaction in the biogenesis of long-chain fatty acids. Carries out three functions: biotin carboxyl carrier protein, biotin carboxylase and carboxyltransferase.
Specifications
| Phospho-Acetyl-CoA Carboxylase (Ser80) | |
| Polyclonal | |
| Unconjugated | |
| ACACA | |
| ACAC, ACACA, ACC-alpha, ACCA, Acetyl-CoA carboxylase 1, EC 6.3.4.14, EC 6.4.1.2, ACC1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 107476, 31, 60581 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13085, Q5SWU9, P11497 | |
| ACACA | |
| A synthetic phosphopeptide derived from human ACC1 around the phosphorylation site of Ser79 (S-M-SP-G-L) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction