Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Acinus (Ser1180) Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5105962
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105962 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105962 Supplier Thermo Scientific Supplier No. PA5105962

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Acinus only when phosphorylated at Ser1180.

ACINUS (apoptotic chromatin condensation inducer in the nucleus) is an induced of chromatin condensation. ACINUS is cleaved by caspase-3 and an additional unknown protease generating a small active peptide, p17, which causes chromatin condensation in vitro when it is added to purified nuclei. ACINUS also induces apoptotic chromatin condensation in cells. ACINUS is ubiquitously expressed. Three different spliced forms of Acinus have been identified in human and mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Acinus (Ser1180)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACIN1
Gene Accession No. Q9JIX8, Q9UKV3
Gene Alias 2610036I19Rik; 2610510L13Rik; Acin1; ACINUS; acinusL; acinusS; Acn; apoptotic chromatin condensation inducer 1; apoptotic chromatin condensation inducer in the nucleus; C79325; DKFZp667N107; fSAP152; functional spliceosome-associated protein 152; KIAA0670; mKIAA0670
Gene Symbols ACIN1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ACIN1(Accession Q9UKV3), corresponding to amino acid residues around phosphorylated Ser1180.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22985, 305884, 56215
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.