Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-ACK1 (Tyr857, Tyr858) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564733
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564733 Supplier Invitrogen™ Supplier No. PA564733
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-ACK1 (Tyr857, Tyr858) Polyclonal Antibody detects endogenous levels of ACK1 only when phosphorylated at Tyr857, Tyr858.

TNK2 (ACK) (activated Cdc42-associated kinase) is a non-receptor tyrosine kinase that signals downstream of Cdc42. TNK2 is known to be involved in tumorigenesis and metastasis, particularly in prostate cancer. Downstream effector of CDC42 which mediates CDC42-dependent cell migration via phosphorylation of BCAR1. May be involved both in adult synaptic function and plasticity and in brain development. Activates AKT1 by phosphorylating it on 'Tyr-176'. Phosphorylates AR on 'Tyr-267' and 'Tyr-363' thereby promoting its recruitment to androgen-responsive enhancers (AREs). Phosphorylates WWOX on 'Tyr-287'. Phosphorylates MCF2, thereby enhancing its activity as a guanine nucleotide exchange factor (GEF) toward Rho family proteins. Contributes to the control of AXL receptor levels. Confers metastatic properties on cancer cells and promotes tumor growth by negatively regulating tumor suppressor such as WWOX and positively regulating pro-survival factors such as AKT1 and AR. Phosphorylates WASP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-ACK1 (Tyr857, Tyr858)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TNK2
Gene Accession No. Q07912
Gene Alias ACK; ACK1; Ack-1; Activated CDC42 kinase 1; activated Cdc42-associated kinase 1; activated p21cdc42Hs kinase; Cdc42 GTPase-inhibiting protein; Cdgip; Non-receptor protein tyrosine kinase Ack; p21cdc42Hs; P21cdc42Hs kinase; Pyk1; Tnk2; tnk2 {ECO:0000250; tyrosine kinase non receptor 2; tyrosine kinase non-receptor protein 2; tyrosine kinase, non-receptor, 2; UniProtKB:Q07912}
Gene Symbols TNK2
Host Species Rabbit
Immunogen A synthesized peptide derived from human Ack1 (Phospho-Tyr857+Tyr858).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10188
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.