Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Aconitase 1 (Ser711) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114590
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114590 Supplier Invitrogen™ Supplier No. PA5114590
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of IREB1 only when phosphorylated at Ser711.

Aconitase 1 (ACO1) also known as iron regulatory element binding protein 1 (IREB1), is a cytosolic protein which binds to iron-responsive elements (IREs). It plays a central role in cellular iron homeostasis. It was also shown to have aconitase activity, and hence grouped with the aconitase family of enzymes.Aconitase 1, also known as iron regulatory element binding protein 1 (IREB1), is a cytosolic protein which binds to iron-responsive elements (IREs). IREs are stem-loop structures found in the 5' UTR of ferritin mRNA, and in the 3' UTR of transferrin receptor mRNA. The iron-induced binding to the IRE results in repression of translation of ferritin mRNA, and inhibition of degradation of the otherwise rapidly degrading transferrin receptor mRNA. Thus, IREB1 plays a central role in cellular iron homeostasis. It was also shown to have aconitase activity, and hence grouped with the aconitase family of enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Aconitase 1 (Ser711)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ACO1
Gene Accession No. P21399, P28271, Q63270
Gene Alias ACO1; Aco-1; Acon1; Aconitase; aconitase 1; aconitase 1, soluble; aconitate hydratase, cytoplasmic; ACONS; AH; Citrate hydro-lyase; cytoplasmic aconitase; cytoplasmic aconitate hydratase; cytosplasmic aconitase; epididymis luminal protein 60; ferritin repressor protein; FRP; HEL60; Ireb1; Irebp; IRE-BP 1; IREBP1; iron regulatory protein 1; iron-responsive element binding protein 1; iron-responsive element-binding protein 1; IRP1; OTTHUMP00000021176; OTTHUMP00000021177; OTTHUMP00000045233; soluble aconitase
Gene Symbols ACO1
Host Species Rabbit
Immunogen A synthesized peptide derived from human IREB1 around the phosphorylation site of Ser711.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11428, 48, 50655
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.