Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-ACTH (Ser168) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982505
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2505 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2505 Supplier Bioss Supplier No. BS12959R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATCH (adrenocorticotropic hormone) is a hormone which plays a major role in stimulating the adrenal cortex. It is formed through cleavage of the polypeptide precursor proopiomelanocortin (POMC), which also results in several other cleavage products including MSH, ACTH, and beta endorphin. ATCH is secreted from the anterior pituitary in response to the corticotropin-releasing hormone from the hypothalamus. It stimulates the secretion of glucocorticoids like cortisol, but has little control over the stimulation of mineralocorticoids like aldosterone, which is another major hormone of the adrenal cortex.

Specifications

Antigen Phospho-ACTH (Ser168)
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Pomc
Gene Accession No. P01189
Gene Alias ACTH; adrenal corticotropic hormone; Adrenocorticotropic hormone; adrenocorticotropin; alpha-melanocyte stimulating hormone; alpha-melanocyte-stimulating hormone; alphaMSH; Alpha-MSH; BE; Beta-endorphin; Beta-LPH; beta-melanocyte-stimulating hormone; Beta-MSH; CLIP; Corticotropin; Corticotropin-like intermediary peptide; Corticotropin-lipotropin; Gamma-LPH; Gamma-MSH; Lipotropin beta; Lipotropin gamma; LPH; Melanocyte-stimulating hormone alpha; Melanocyte-stimulating hormone beta; Melanotropin alpha; Melanotropin beta; Melanotropin gamma; Met-enkephalin; MSH; NPP; opiomelanocortin prepropeptide; OTTHUMP00000119991; OTTHUMP00000200964; POC; Pomc; Pomc1; Pomc-1; Pomc2; Potential peptide; Precursor of MSH; pro-ACTH-endorphin; proopiomelanocortin; Pro-opiomelanocortin; proopiomelanocortin (adrenocorticotropin/ beta-lipotropin/ alpha-melanocyte stimulating hormone/ beta-melanocyte stimulating hormone/ beta-endorphin); proopiomelanocortin preproprotein; proopiomelanocortin, beta (endorphin, beta); pro-opiomelanocortin-alpha; proopoimelanocortin, beta (endorphin, beta)
Gene Symbols Pomc
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human ACTH around the phosphorylation site of Ser168.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5443
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.