Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-ADAM9 (Thr761) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105334
Description
Antibody detects endogenous levels of ADAM9 only when phosphorylated at Thr761.
ADAM9 is a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This protein interacts with SH3 domain-containing proteins, binds mitotic arrest deficient 2 beta protein, and is also involved in TPA-induced ectodomain shedding of membrane-anchored heparin-binding EGF-like growth factor.
Specifications
Phospho-ADAM9 (Thr761) | |
Polyclonal | |
Unconjugated | |
ADAM9 | |
a disintegrin and metallopeptidase domain 9 (meltrin gamma); a disintegrin and metalloprotease domain 9; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 9 (meltrin gamma); ADAM; ADAM 9; ADAM metallopeptidase domain 9; ADAM metallopeptidase domain 9 (meltrin gamma); adam9; ADAMs; AU020942; Cellular disintegrin-related protein; cone rod dystrophy 9; CORD9; Disintegrin and metalloproteinase domain-containing protein 9; Kiaa0021; MCMP; MDC 9; MDC9; meltrin gamma; meltrin-gamma; metalloendopeptidases; Metalloprotease/disintegrin/cysteine-rich protein 9; mKIAA0021; Mltng; Myeloma cell metalloproteinase; OTTHUMP00000216481; OTTHUMP00000216484; OTTHUMP00000216485 | |
Rabbit | |
Sequential chromatography | |
RUO | |
11502, 290834, 8754 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q13443, Q61072 | |
ADAM9 | |
A synthesized peptide derived from human ADAM9(Accession Q13443), corresponding to amino acid residues around phosphorylated Thr761. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction