Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho AEP (Ser226), Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABN2270100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN2270100 100 μg
ABN227025UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN2270100 Supplier MilliporeSigma Supplier No. ABN2270100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho AEP (Ser226), Cat. No. ABN2270, is a highly specific rabbit polyclonal antibody that targets Asparaginyl endopeptidase (AEP) and has been tested in Immunofluorescence, serine-arginine protein kinase 2 (SRPK2) Assay, and Western Blotting.

Legumain (UniProt: Q99538; also known as EC: 3.4.22.34, Asparaginyl endopeptidase, AEP, Protease cysteine 1, Delta Secretase) is encoded by the LGMN (also known as PRSC1) gene (Gene ID: 5641) in human. AEP is a lysosomal endopeptidase that displays strict specificity for the hydrolysis of asparaginyl bonds and can simultaneously cleave both amyloid precursor protein (APP) and tau. It cleaves aspartyl bonds slowly, especially under acidic conditions. AEP is synthesized with a signal peptide (aa 1-17) and a propeptide (aa 324-433) that are cleaved off in the mature form. It exists a homodimer before removal of the propeptide and after autocatalytic processing exists as a monomer. In its zymogen form, the uncleaved propeptide blocks access to the active site. AEP is phosphorylated on serine 226 by serine-arginine protein kinase 2 (SRPK2), which accelerates its autocatalytic cleavage and enhances its enzymatic activity. The active form translocates to cytoplasm. In Alzheimer s disease brains AEP is reported to be highly phosphorylated. Depletion of AEP is shown to reduce amyloid peptide production and senile plaque formation in 5XFAD mouse brain that leads to restoration of synaptic activity and cognitive function. Three isoforms of AEP have been described that are produced by alternative splicing. (Ref.: Wang Z-H., et al. (2017). Mol. Cell 67(5); 812-825; Zhang, Z et al (2015). Nat. Commun. 6; p. 9762).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho AEP (Ser226)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blotting Analysis: A 1:2,000 dilution from a representative lot detected phospho AEP (Ser226) in lysates from HEK293 cells co-transfected mammalian GST-tagged (mGST) AEP and wild-type (WT) SRPK2. (Courtesy of Keqiang Ye, Ph.D., Professor, Emory University School of Medicine, Department of Pathology and Laboratory Medicine).Immunofluorescence Analysis: A representative lot detected phospho AEP (Ser226) in Immunofluorescence applications (Wang, Z.H., et. al. (2017). Mol Cell. 67(5):812-825).Kinase Assay: A representative lot detected phospho AEP (Ser226) in HEK293 cells treated with SRPK2 and in vitro kinase assays (Wang, Z.H., et. al. (2017). Mol Cell. 67(5):812-825).
Formulation Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide.
Gene Alias Legumain;Asparaginyl endopeptidase;Protease cysteine 1;Delta Secretase
Gene Symbols LGMN;PRSC1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 13 amino acids surrounding phosphoserine 226 of human Asparaginyl endopeptidase (AEP).
Molecular Weight of Antigen ∽73 kDa observed; 49.41 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_005597
Test Specificity This rabbit polyclonal antibody specifically detects Asparaginyl endopeptidase phosphorylated on Serine 226.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.