Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Afadin (Ser1721) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985124
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5124 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5124 Supplier Bioss Supplier No. BS3072R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AFDN belongs to an adhesion system, probably together with the E-cadherin-catenin system, which plays a role in the organization of homotypic, interneuronal and heterotypic cell-cell adherens junctions (AJs). Nectin- and actin-filament-binding protein that connects nectin to the actin cytoskeleton.

Specifications

Antigen Phospho-Afadin (Ser1721)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Afdn
Gene Accession No. P55196
Gene Alias 5033403D15Rik; AF6; Af-6; Afadin; Afadin adherens junction formation factor; afadin, adherens junction formation factor; AFDN; ALL1-fused gene from chromosome 6 protein; FLJ34371; Gm314; I-afadin; MLL-AF6; Mllt4; myeloid/lymphoid or mixed lineage-leukemia translocation to 4 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 4; myeloid/lymphoid or mixed-lineage leukemia 4; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 4; Protein Af-6; RP3-431P23.3; RP3-470B24.4; S-afadin
Gene Symbols Afdn
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human Afadin around the phosphorylation site of Ser1721 [VL (p-S)PD].
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4301
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.