Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Afadin (Tyr1230) Polyclonal Antibody

Phospho-Afadin (Tyr1230) Polyclonal Antibody
Supplier: Thermo Scientific PA5105593
Description
Phospho-Afadin (Tyr1230) Polyclonal Antibody for Western Blot
Specifications
| Phospho-Afadin (Tyr1230) | |
| Polyclonal | |
| Unconjugated | |
| Afdn | |
| 5033403D15Rik; Af6; Af-6; Afadin; Afadin adherens junction formation factor; afadin, adherens junction formation factor; afadin; afadin, adherens junction formation factor; AFDN; ALL1-fused gene from chromosome 6 protein; FLJ34371; Gm314; I-afadin; MLL-AF6; MLLT4; myeloid/lymphoid or mixed lineage-leukemia translocation to 4 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 4; myeloid/lymphoid or mixed-lineage leukemia 4; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 4; protein Af-6; RP3-431P23.3; RP3-470B24.4; S-afadin | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 17356, 26955, 4301 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35889, P55196, Q9QZQ1 | |
| Afdn | |
| A synthesized peptide derived from human AFDN(Accession P55196), corresponding to amino acid residues around phosphorylated Tyr1230. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction