Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-AHNAK (Ser5782) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564530
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564530 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564530 Supplier Invitrogen™ Supplier No. PA564530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-AHNAK (Ser5782) Polyclonal Antibody detects endogenous levels of AHNAK only when phosphorylated at Ser5782.

AHNAK1 (Desmoyokin) is a large (700 kDa) scaffold protein that translocates to the plasma membrane after an increas of extracellular calcium level or upon proteinkinase C activation and regulates extracellular calcium influx mediated by L-type Ca2+ channels. AHNAK1 has been implicated in diverse signal transduction proceses affecting cell differentiation and proliferation. In response to calcium-dependent intercellular contacts AHNAK1 forms multimeric complexes in the plasma membrane, connected with actin and annexin 2/S100A10 assemblies and is thus involved in organization of the plasma membrane architecture. In epithelial cells, AHNAK1 is localized in cytoplasm or is membrane-associated, but in cells of nonepithelial origin AHNAK1 is predominantly nuclear; it has a weak DNA-binding activity and associates with the DNA-ligase IV-XRCC4 complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-AHNAK (Ser5782)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AHNAK
Gene Accession No. Q09666
Gene Alias AHNAK; AHNAK 1; AHNAK nucleoprotein; AHNAK nucleoprotein (desmoyokin); AHNAK-related; AHNAKRS; Desmoyokin; DY6; MGC5395; Neuroblast differentiation-associated protein AHNAK; PM227; RGD1308572; UV118
Gene Symbols AHNAK
Host Species Rabbit
Immunogen A synthesized peptide derived from human Desmoyokin (Phospho-Ser5782).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66395, 79026
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.