Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Phospho-Akt (S473) Pan Specific Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 25 publications
Supplier: R&D Systems AF887SP
This item is not returnable.
View return policy
Description
AKT Polyclonal specifically detects AKT in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
| AKT (p Ser473) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AKT1 | |
| Phosphopeptide containing human, mouse, and rat Akt1 S473 site | |
| 25 μg | |
| Primary | |
| Detects human, mouse and rat Akt1, Akt2 and Akt3, when phosphorylated at S473, S474 and S472, respectively. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 5-15 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB alpha, PKB beta, PKB gamma, PKB-ALPHA, PRKBA, RAC, RAC-alpha, RAC-beta, RAC-gamma | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 207 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction