Learn More
Phospho-AKT (Ser473) Monoclonal Antibody (17F6.B11), Rockland™

Mouse Monoclonal Antibody
$178.35 - $677.73
Specifications
| Antigen | Phospho-AKT (Ser473) |
|---|---|
| Clone | 17F6.B11 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-194-3788
![]()
|
Rockland Immunochemicals
200301268 |
100 μg |
Each for $677.73
|
|
|||||
50-199-0513
![]()
|
Rockland Immunochemicals
200301268S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This phospho specific monoclonal antibody is specific for phosphorylated human and mouse AKT protein at S473. A BLAST analysis was used to suggest cross-reactivity with AKT pS473 from human, mouse, rat and chimpanzee sources based on 100% homology with the immunizing sequence. Cross-reactivity with AKT from other sources has not been determined. Cross-reactivity with AKT2 and AKT3 has not been determined.
AKT also known as protein kinase B (PKB) or RAS-alpha, is an ubiquitous serine/threonine kinase that plays an important role in diverse biological responses such as regulation of metabolism, cell survival and growth by phosphorylating multiple proteins. This protein kinase is activated by insulin, PI3K, IGF1 and various other growth and survival factors. Akt promotes cell survival by inhibiting apoptosis through phosphorylation and inactivation of several targets, including forkhead transcription factors, and caspase-9. The AKT pathway is a major target for cancer drug discovery.Specifications
| Phospho-AKT (Ser473) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| Akt; Akt kinase; AKT serine/threonine kinase 1; AKT/PKB; AKT1; AKT1 kinase; AKT1m; Akt1m protein; Akt1-PA; Akt1-PB; Akt1-PC; Akt1-PD; Akt1-PE; AKT2; AKT3; CAKT; C-AKT; CG4006; CG4006 gene product from transcript CG4006-RE; CG4006-PA; CG4006-PB; CG4006-PC; CG4006-PD; CG4006-PE; CWS6; DAkt; D-Akt; dAkt kinase; dAKT/dPKB; dAkt/PKB; DAkt1; DAKT1/PKB; Dmel\CG4006; Dmel_CG4006; dPKB; DRAC-PK; DRAC-PK66; DRAC-PK85; kinase Akt1; l(3)04226; l(3)89Bq; MGC99656; murine thymoma viral (v-akt) oncogene homolog 1; pAkt; p-Akt; Phospho-AKT; Phospho-PKB; PKB; PKB Akt; PKB alpha; PKB beta; PKB gamma; PKB/AKT; PKB/dAKT; PKBalpha; PKB-ALPHA; PKBG; PRKBA; Protein kinase B; Protein kinase B alpha; protein kinase B-alpha; Proto-oncogene c-Akt; Rac; rac protein kinase alpha; RAC protein kinase alpha RAC-PK alpha; RAC serine/threonine-protein kinase; RAC-ALPHA; RAC-alpha serine/threonine-protein kinase; RacPK; RAC-PK-alpha; RAC-PK-beta; RAC-PK-gamma; related to A and C kinases; related to PKA to PKC protein kinases; related to the A and C kinases; serine/threonine protein kinase; serine-threonine protein kinase; STK-2; Thymoma viral proto-oncogene; thymoma viral proto-oncogene 1; v-akt murine thymoma viral oncogene homolog 1; v-akt murine thymoma viral oncogene-like protein 1 | |
| AKT1 | |
| IgG1 κ | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| AKT1 |
| 17F6.B11 | |
| ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Monkey, Mouse, Rat | |
| P31749, P31750, P47196 | |
| 11651, 207, 24185 | |
| Anti-AKT pS473 (MOUSE) Monoclonal Antibody was produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding S473 of human AKT1 protein, followed by hybridoma development. | |
| Primary | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.