Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Akt1 (Ser473) Recombinant Rabbit Monoclonal Antibody (AktS473-B9), PE, Invitrogen™
GREENER_CHOICE

Catalog No. PIMA537063
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537063 100 Tests
PIMA537323 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537063 Supplier Invitrogen™ Supplier No. MA537063
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

AKT1 (PKB alpha) is a serine/threonine kinase that regulates cell survival. The activated enzyme inhibits apoptosis and stimulates cell cycle progression by phosphorylating numerous targets in various cell types, including cancer cells. This protein kinase is activated by insulin, PI3K, IGF1 and various other growth and survival factors. Akt promotes cell survival by inhibiting apoptosis through phosphorylation and inactivation of several targets, including forkhead transcription factors, and caspase-9. The AKT pathway is a major target for cancer drug discovery.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Akt1 (Ser473)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone AktS473-B9
Conjugate PE
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene AKT1
Gene Accession No. P31749, P31750
Gene Alias AKT; Akt kinase; AKT serine/threonine kinase 1; Akt/PKB; akt1; AKT1 kinase; AKT1m; Akt1m protein; Akt1-PA; Akt1-PB; Akt1-PC; Akt1-PD; Akt1-PE; CAKT; C-AKT; CG4006; CG4006-PA; CG4006-PB; CG4006-PC; CG4006-PD; CG4006-PE; CWS6; dAkt; D-Akt; dAkt kinase; dAKT/dPKB; dAkt/PKB; DAKT1; DAKT1/PKB; Dmel\CG4006; Dmel_CG4006; dPKB; DRAC-PK; DRAC-PK66; DRAC-PK85; l(3)04226; l(3)89Bq; murine thymoma viral (v-akt) oncogene homolog 1; pAkt; P-AKT; PKB; PKB alpha; PKB beta; PKB gamma; PKB/AKT; PKB/dAKT; PKBalpha; PKB-ALPHA; PKBG; PRKBA; protein kinase B; Protein kinase B alpha; protein kinase B-alpha; proto-oncogene c-Akt; RAC; rac protein kinase alpha; RAC protein kinase alpha RAC-PK alpha; RAC serine/threonine protein kinase; RAC serine/threonine-protein kinase; RAC-ALPHA; RAC-ALPHA SERINE/THREONINE-PROTEIN KINASE; RacPK; RAC-PK-alpha; RAC-PK-beta; RAC-PK-gamma; related to A and C kinases; related to PKA to PKC protein kinases; related to the A and C kinases; SD10374p; serine/threonine protein kinase; serine-threonine protein kinase; Thymoma viral proto-oncogene; thymoma viral proto-oncogene 1; v-akt murine thymoma viral oncogene homolog 1; V-AKT MURINE THYMOMA VIRAL ONCOGENE-LIKE PROTEIN 1
Gene Symbols AKT1
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Ser473 of human phospho Akt1.
Purification Method Protein A/G
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11651, 207
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.