Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-alpha 1 Sodium Potassium ATPase (Tyr260) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983934
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3934 Supplier Bioss Supplier No. BS15967R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene belongs to the family of P-type cation transport ATPases, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The catalytic subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes an alpha 1 subunit. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen Phospho-alpha 1 Sodium Potassium ATPase (Tyr260)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ATP1A1
Gene Accession No. P05023
Gene Alias (Na+ + K+)-ATPase; (Na+, K+)-ATPase alpha-subunit; ATP1A1; atp1a1-a; atp1a1a.4; atp1a1-b; ATP1B; Atpa-1; ATPase Na+/K+ transporting subunit alpha 1; ATPase, Na+/K+ transporting, alpha 1 polypeptide; ATPase, Na+K+ transporting, alpha 1; BC010319; BOS_3049; K-ATPase; MGC1798; Na(+)/K(+) ATPase alpha-1 subunit; Na, K-ATPase, alpha-A catalytic polypeptide; Na,K-ATPase alpha-1 subunit; Na,K-ATPase catalytic subunit alpha-A protein; Na/K ATPase alpha 1 isoform; Na/K ATPase alpha 1 subunit; Na+/K+ ATPase; Na+/K+ ATPase 1; Na+/K+ ATPase Alpha; Na+/K+ -ATPase alpha 1 subunit; Na+/K+ transporting alpha 1 polypeptide; Na+K+ ATPase; Na+K+ATPase; Nkaa1b; RP4-655J12.1; Sodium Potassium ATPase; sodium pump 1; Sodium pump subunit alpha-1; sodium/potassium-ATPase alpha-subunit 1; sodium/potassium-transporting ATPase alpha-1 chain; Sodium/potassium-transporting ATPase subunit alpha-1; sodium-potassium ATPase catalytic subunit alpha-1; sodium-potassium-ATPase, alpha 1 polypeptide; unnamed protein product
Gene Symbols ATP1A1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human alpha 1 Sodium Potassium ATPase around the phosphorylation site of Tyr260.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 476
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.