Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-alpha Adducin (Ser726) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5104878

Description
Phospho-alpha Adducin (Ser726) Polyclonal antibody specifically detects Phospho-alpha Adducin (Ser726) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Specifications
Phospho-alpha Adducin (Ser726) | |
Polyclonal | |
Unconjugated | |
ADD1 | |
Add1; ADD2; ADDA; ADDB; adducin 1; adducin 1 (alpha); Adducin beta; Adducin1; Adducin-1; Adducin-alpha; Adducin-beta; AI256389; Alpha adducin; alpha-ADD; alpha-adducin; beta Adducin; Beta-adducin; erythrocyte adducin alpha subunit; Erythrocyte adducin subunit alpha | |
A synthesized peptide derived from human ADD1 around the phosphorylation site of Ser726. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P35611, Q63028, Q9QYC0 | |
Rabbit | |
sequential chromatography | |
RUO | |
11518, 118, 24170 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction