Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-alpha B Crystallin (Ser45) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982510
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2510 Supplier Bioss Supplier No. BS12970R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lens proteins consist almost entirely of crystallins (about 95%). Crystallins are also found vertebrate skeletal muscle tissue. In the lens, their structural function is to assist in maintaining the proper refractive index of the lens. The mammalian lens contains 3 major classes of crystallins: alpha, beta, and gamma. Alpha-crystallin is the largest of the crystallins and is composed of 2 primary gene products-alpha-A and alpha-B. There are at least 5 different proteins comprising the beta-crystallins. The gamma-crystallins are monomeric, but there are at least 5 gamma crystallins identified in bovine and rat lens. Alpha-Crystallin comprises 40% of total lens protein composition. In addition to maintaining proper refractive index, it also functions in a chaperone like manner by preventing the formation of aggregates possibly leading to cataract formation. It is believed that the phosphorylated states of the alpha-crystallin occur in response to cellular stress and may serve a structural control function and play a role in protein maintenance. Alpha-B crystallin has been linked to Alexander′s disease where it accumulates in brain cells of those afflicted.

Specifications

Antigen Phospho-alpha B Crystallin (Ser45)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Cryab
Gene Accession No. P02511
Gene Alias AACRYA; alpha B-crystallin; Alpha(B)-crystallin; alpha-B-crystallin; Alpha-crystallin B chain; alpha-crystallin B chain-like protein; BOS_14343; CMD1II; CRYA2; Crya-2; Cryab; CryAB antibody; crystallin alpha B; crystallin, alpha 2; crystallin, alpha B; crystallin, alpha polypeptide 2; CTPP2; CTRCT16; epididymis secretory protein Li 101; Heat shock protein beta-5; heat shock protein CryAB; heat shock protein HspB5; heat-shock 20 kD like-protein; HEL-S-101; HSPB5; MFM2; P23; Renal carcinoma antigen NY-REN-27; Rosenthal fiber component
Gene Symbols Cryab
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human alpha B Crystallin around the phosphorylation site of Ser45.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1410
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.