Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-AMPK alpha-1,2 (Thr183, Thr172) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110151
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110151 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110151 Supplier Invitrogen™ Supplier No. PA5110151
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-AMPK alpha-1,2 (Thr183, Thr172) Polyclonal Antibody for ELISA, IHC (P), IP, Western Blot

The protein encoded by this gene is a catalytic subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. Studies of the mouse counterpart suggest that this catalytic subunit may control whole-body insulin sensitivity and is necessary for maintaining myocardial energy homeostasis during ischemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-AMPK alpha-1,2 (Thr183, Thr172)
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene PRKAA2
Gene Accession No. P54645, P54646, Q09137, Q13131, Q5EG47, Q8BRK8
Gene Alias 2310008I11Rik; 5'-AMP-activated protein kinase alpha-1 catalytic subunit; 5'-AMP-activated protein kinase alpha-2 catalytic subunit; 5'-AMP-activated protein kinase catalytic alpha-2 subunit; 5'-AMP-activated protein kinase catalytic subunit alpha-1; 5'-AMP-activated protein kinase catalytic subunit alpha-2; 5'-AMP-activated protein kinase, catalytic alpha-1 chain; 5'-AMP-activated protein kinase, catalytic alpha-2 chain; 63 kDa subunit; A830082D05; AAPK2; ACACA kinase; acetyl-CoA carboxylase kinase; AI194361; AI450832; AL024255; AMP -activate kinase alpha 1 subunit; AMP-activated protein kinase; AMP-activated protein kinase alpha 2; AMP-activated protein kinase alpha-2 subunit variant 2; AMP-activated protein kinase alpha-2 subunit variant 3; AMP-activated protein kinase alpha-2 variant B; AMP-activated protein kinase, alpha 1 catalytic subunit; AMP-activated protein kinase, catalytic, alpha-1; AMPK; AMPK 63 kDa subunit; AMPK alpha 1; AMPK alpha 2; AMPK alpha-1 chain; AMPK alpha-2 chain; AMPK subunit alpha-1; AMPK subunit alpha-2; AMPK1; AMPK2; AMPKa1; AMPKa2; AMPK-activated protein kinase alpha-2 subunit; AMPKalpha1; AMPK-alpha-1 chain; AMPKalpha2; AMPK-alpha-2 chain; C130083N04Rik; HMGCR kinase; hydroxymethylglutaryl-CoA reductase kinase; pAMPK; PRKAA; Prkaa1; Prkaa2; protein kinase; protein kinase AMP-activated alpha 2 catalytic subunit; protein kinase AMP-activated catalytic subunit alpha 1; protein kinase AMP-activated catalytic subunit alpha 2; protein kinase, AMP-activated, alpha 1 catalytic subunit; protein kinase, AMP-activated, alpha 2 catalytic subunit; tau-protein kinase PRKAA1
Gene Symbols PRKAA1, PRKAA2
Host Species Rabbit
Immunogen A phospho specific peptide corresponding to residues surrounding T172 of human AMPK alpha.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105787, 108079, 5562, 5563, 65248, 78975
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.