Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-AMPK alpha-1 (Ser496, Ser502) Rabbit anti-Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501985093
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5093 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5093 Supplier Bioss Supplier No. BS3025R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.

Specifications

Antigen Phospho-AMPK alpha-1 (Ser496, Ser502)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Prkaa1
Gene Accession No. P54645, Q5EG47
Gene Alias 5′-AMP-activated protein kinase alpha-1 catalytic subunit; 5′-AMP-activated protein kinase catalytic subunit alpha-1; 5′-AMP-activated protein kinase, catalytic alpha-1 chain; 63 kDa subunit; AAPK2; ACACA kinase; Acetyl-CoA carboxylase kinase; activated protein kinase; AI194361; AI450832; AL024255; AMP -activate kinase alpha 1 subunit; AMP activated protein kinase; AMP-activated protein kinase, alpha 1 catalytic subunit; AMP-activated protein kinase, catalytic, alpha-1; AMPK; AMPK 63 kDa subunit; AMPK alpha 1; AMPK alpha-1 chain; AMPK subunit alpha-1; AMPK subunit pAMPK; AMPK1; AMPK2; AMPKa1; AMPKalpha1; AMPK-alpha-2 chain; BOS_19826; C130083N04Rik; HMGCR kinase; Hydroxymethylglutaryl-CoA reductase kinase; MGC33776; MGC57364; pAMPK; PRKAA; PRKAA1; PRKAA2; protein kinase AMP-activated catalytic subunit alpha 1; protein kinase, AMP-activated, alpha 1 catalytic subunit; Tau-protein kinase PRKAA1
Gene Symbols Prkaa1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from mouse AMPK alpha-1 around the phosphorylation site of Ser496+Ser502.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105787, 65248
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.