Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ phospho-AMPK alpha-1 (Thr479), Rabbit, Unlabeled, Polyclonal,

Catalog No. ABS981 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABS981 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS981 Supplier MilliporeSigma™ Supplier No. ABS981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

5&prime-AMP-activated protein kinase catalytic subunit alpha-1 (EC 2.7.11.1; EC 2.7.11.26; EC 2.7.11.27; EC 2.7.11.31; UniProt Q13131; also known as ACACA kinase, Acetyl-CoA carboxylase kinase, AMPK subunit alpha-1, AMPKa1, HMGCR kinase, Hydroxymethylglutaryl-CoA reductase kinase, Tau-protein kinase PRKAA1) is encoded by the PRKAA1 (also known as AMPK1) gene (Gene ID 5562) in human. The yeast sucrose non-fermenting 1 (SNF1) and the AMP-activated protein kinase (AMPK) in drosphila and mammals are orthologous heterotrimeric serine-threonine kinases, each consisting of a catalytic α subunit and regulatory β and γ subunits, that sense decreased cellular energy levels when the AMP to ATP ratio rises due to a high rate of metabolism or decreased nutrient availability. AMP binding to the γ subunit results in an enhanced phosphorylation of the catalytic α subunit on an activation loop threonine and AMPK allosteric activation. Activated AMPK phosphorylates downstream targets, leading to energy conservation and inhibition of ATP consumption. For example, the high rate of ATP use in contracting muscle causes AMPK-mediated phosphorylation/inactivation of acetyl-CoA carboxylase (ACC), thereby increasing fatty acid oxidation and restoring cellular energy. AMPK functions as a positive regulator of autophagy by phosphorylating the protein kinase ULK1 that initiates autophagy as well as by inhibiting the mammalian TOR (mTOR) pathway activity via phosphorylating mTOR-binding partner raptor. In mammals, there exist two different α (encoded by the PRKAA1 and PRKAA2 gene), two different β (encoded by the PRKAB1 and PRKAB2 gene), and three different γ (encoded by the PRKAG1, PRKAG2, and PRKAG3 gene) subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-AMPK alpha-1 (Thr479)
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene PRKAA1, AMPK1
Gene Accession No. Q13131
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the AMPK alpha-1 phosphorylation site sequence containing the phosphorylated target threonine.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) AAC52355
Test Specificity The Thr479 numbering is based on the rat AMPK alpha-1 sequence shown in Fig 2A of Suzuki, T., et al. (2013). Mol. Cell. 50(3):407-419, which was cited for as Thr479 phosphorylation in subsequent publications (Coughlan, K.A., et al. (2015). PLoS One. 10(5):e0127388; Hawley, S.A., et al. (2014). Biochem. J. 459(2):275-287). The target threonine corresponds to Thr490 of the human/mouse/rat AMPK alpha-1 sequences published by UniProt (Q13131-1/Q5EG47/P54645) or Thr505 of human spliced isoform 2 (UniProt.Q13131-2). The target site sequence is not well conserved between AMPK alpha-1 (-SGTA(pT)PQR-) and AMPK alpha-2 (-SGSS(pT)PQR-).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.