Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-AMPK-alpha (Thr172) Polyclonal Antibody, Abbomax

Catalog No. 502204462
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4462 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4462 Supplier Abbomax Inc Supplier No. 620120
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: EGF-stimulated A431 Cellular location: Cytoplasm.

Phospho-AMPK-alpha (Thr172) Polyclonal antibody specifically detects Phospho-AMPK-alpha (Thr172) in Bovine, Canine, Chicken, Human, Insect, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Phospho-AMPK-alpha (Thr172)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation 25mM tris with proprietary stabilizer and 0.01% sodium azide
Gene PRKAA2
Gene Accession No. P54646, Q09137, Q8BRK8
Gene Alias 2310008I11Rik; 5'-AMP-activated protein kinase catalytic subunit alpha-2; 5'-AMP-activated protein kinase, catalytic alpha-2 chain; A830082D05; ACACA kinase; Acetyl-CoA carboxylase kinase; AMP-activated protein kinase; AMP-activated protein kinase alpha-2 subunit variant 2; AMP-activated protein kinase alpha-2 subunit variant 3; AMP-activated protein kinase alpha-2 variant B; AMPK; AMPK alpha 2; AMPK alpha-2 chain; AMPK subunit alpha-2; AMPK2; AMPKa2; AMPKalpha2; AMPK-alpha-2 chain; EC 2.7.11.1; HMGCR kinase; hydroxymethylglutaryl-CoA reductase kinase; PRKAA; Prkaa2; protein kinase AMP-activated catalytic subunit alpha 2; protein kinase, AMP-activated, alpha 2 catalytic subunit
Host Species Rabbit
Immunogen A synthetic peptide surrounding to the epitope -LRTSC- with a phosphorylation site at Thr 172 of AMPK alpha protein from human origin
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108079, 429110, 489571, 538954, 5563, 78975
Target Species Bovine, Canine, Chicken, Human, Insect, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.