Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-AMPK beta 1 (Ser108) Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS3026R

Description
AMPK beta 1 is a regulatory subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This subunit may be a positive regulator of AMPK activity. The myristoylation and phosphorylation of this subunit have been shown to affect the enzyme activity and cellular localization of AMPK. This subunit may also serve as an adaptor molecule mediating the association of the AMPK complex.Specifications
Phospho-AMPK beta 1 (Ser108) | |
Polyclonal | |
Unconjugated | |
PRKAB1 | |
1300015D22Rik; 5′-AMP-activated protein kinase 40 kDa subunit; 5-AMP-activated protein kinase beta subunit; 5′-AMP-activated protein kinase beta-1 subunit; 5′-AMP-activated protein kinase subunit beta-1; 5′-AMP-activated protein kinase subunit beta-1-like protein; 5′-AMP-activated protein kinase, beta subunit; AAKB1; AMP-activated protein kinase beta subunit; AMPK; AMPK beta 1; AMPK beta -1 chain; AMPK beta-1 chain; AMPK subunit beta-1; AMPKb; AMPK-beta1; AU021155; E430008F22; HAMPKb; kinase AMPK-beta1; kinase pAMPK; pAMPK; PRKAB1; protein kinase AMP-activated non-catalytic subunit beta 1; protein kinase, AMP-activated, beta 1 non-catalytic subunit; protein kinase, AMP-activated, noncatalytic, beta-1 | |
Rabbit | |
Protein A | |
RUO | |
5564 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9Y478 | |
PRKAB1 | |
KLH conjugated synthetic phosphopeptide derived from mouse AMPK beta 1 around the phosphorylation site of Ser108. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction