Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-AMPK beta-1 (Ser181) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105772
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105772 Supplier Invitrogen™ Supplier No. PA5105772
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of AMPK beta 1 only when phosphorylated at Ser181.

AMPK beta 1 is a regulatory subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This subunit may be a positive regulator of AMPK activity. The myristoylation and phosphorylation of this subunit have been shown to affect the enzyme activity and cellular localization of AMPK. This subunit may also serve as an adaptor molecule mediating the association of the AMPK complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-AMPK beta-1 (Ser181)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PRKAB1
Gene Accession No. P80386, Q9R078, Q9Y478
Gene Alias 1300015D22Rik; 5'-AMP-activated protein kinase 40 kDa subunit; 5-AMP-activated protein kinase beta subunit; 5'-AMP-activated protein kinase beta-1 subunit; 5'-AMP-activated protein kinase subunit beta-1; 5'-AMP-activated protein kinase subunit beta-1-like protein; 5'-AMP-activated protein kinase, beta subunit; AAKB1; AMP-activated protein kinase beta subunit; AMPK; AMPK beta 1; AMPK beta -1 chain; AMPK beta-1 chain; AMPK subunit beta-1; AMPKb; AMPK-beta1; AU021155; E430008F22; HAMPKb; kinase AMPK-beta1; kinase pAMPK; pAMPK; Prkab1; protein kinase AMP-activated non-catalytic subunit beta 1; protein kinase, AMP-activated, beta 1 non-catalytic subunit; protein kinase, AMP-activated, noncatalytic, beta-1
Gene Symbols PRKAB1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PRKAB1(Accession Q9Y478), corresponding to amino acid residues around phosphorylated Ser181.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19079, 5564, 83803
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.