Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-APLF (Ser116) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105966
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105966 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105966 Supplier Invitrogen™ Supplier No. PA5105966
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of APLF only when phosphorylated at Ser116.

C2ORF13 is a component of the cellular response to chromosomal DNA single- and double-strand breaks.[supplied by OMIM, Mar 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-APLF (Ser116)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene APLF
Gene Accession No. Q8IW19, Q9D842
Gene Alias 2010301N04Rik; AI452191; APFL; APLF; aprataxin- and PNK-like factor; Aprataxin and PNK-like factor; aprataxin and PNKP like factor; apurinic-apyrimidinic endonuclease APLF; C2orf13; PALF; PNK and APTX-like FHA domain-containing protein; PNK and APTX-like FHA protein; RGD1565557; XIP1; XRCC1-interacting protein 1; ZCCHH1; zinc finger, CX5CX6HX5H motif containing 1
Gene Symbols APLF
Host Species Rabbit
Immunogen A synthesized peptide derived from human APLF(Accession Q8IW19), corresponding to amino acid residues around phosphorylated Ser116.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200558, 500247, 72103
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.