Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-AQP2 (Ser264) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986396
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6396 Supplier Bioss Supplier No. BS5185R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aquaporin 2 (AQP2) is a hormonally regulated water channel located in the renal collecting duct. Mutations in the AQP2 gene cause hereditary nephrogenic diabetes insipidus in humans. A vasopressin induced cAMP increase results in the phosphorylation of AQP2 at serine-256 and its translocation from the intracellular vesicles to the apical membrane of principal cells. Recently, serine-261 has been identified as a novel phosphorylation site on AQP2 and levels of phosphorylated S261 have been shown to decrease with vasopressin treatment suggesting its involvement in vasopressin-dependent AQP2 trafficking.

Specifications

Antigen Phospho-AQP2 (Ser264)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Aqp2
Gene Accession No. P41181
Gene Alias ADH water channel; AQP 2; AQP CD; Aqp2; AQP-2; AQPCD; AQP-CD; aquaporin 2; aquaporin 2 (collecting duct); Aquaporin CD; Aquaporin2; Aquaporin-2; Aquaporin-CD; Aquaporine 2; Collecting duct water channel protein; cph; jpk; MGC34501; Water channel protein for renal collecting duct; water-channel aquaporin 2; WCH CD; WCHCD; WCH-CD
Gene Symbols Aqp2
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human AQP2 around the phosphorylation site of Ser264.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 359
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.