Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-ARHGAP31 (Thr789) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105977
Description
Antibody detects endogenous levels of CdGAP only when phosphorylated at Thr789.
This gene encodes a GTPase-activating protein (GAP). A variety of cellular processes are regulated by Rho GTPases which cycle between an inactive form bound to GDP and an active form bound to GTP. This cycling between inactive and active forms is regulated by guanine nucleotide exchange factors and GAPs. The encoded protein is a GAP shown to regulate two GTPases involved in protein trafficking and cell growth.
Specifications
Phospho-ARHGAP31 (Thr789) | |
Polyclonal | |
Unconjugated | |
ARHGAP31 | |
5830477L08Rik; AOS1; ARHGAP31; AU041750; Cdc42 GTPase-activating protein; CDGAP; D330026I07Rik; KIAA1204; Rho GTPase activating protein 31; rho GTPase-activating protein 31 | |
Rabbit | |
sequential chromatography | |
RUO | |
12549, 288093, 57514 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A6X8Z5, Q2M1Z3 | |
ARHGAP31 | |
A synthesized peptide derived from human ARHGAP31(Accession Q2M1Z3), corresponding to amino acid residues around phosphorylated Thr789. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction