Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-ARHGAP42 (Tyr792) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105370
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105370 Supplier Invitrogen™ Supplier No. PA5105370
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of ARHGAP42 only when phosphorylated at Tyr792.

This gene encodes a Rho GTPase-activating protein (RhoGAP), and member of the GRAF or BAR-PH family of proteins. Expression of this gene is enriched in vascular smooth muscle cells and the encoded protein inhibits RhoA activity to regulate vascular tone and control blood pressure. A mutation in the first intron of this gene modulates its expression and is associated with reduced blood pressure in human patients with borderline hypertension.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-ARHGAP42 (Tyr792)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARHGAP42
Gene Accession No. A6NI28, B2RQE8
Gene Alias 9030420J04Rik; Arhgap42; BE136619; Graf3; RGD1564081; Rho GTPase activating protein 42; Rho GTPase-activating protein 10-like; Rho GTPase-activating protein 42; rho-type GTPase-activating protein 42; TMEM133; VRho GTPase activating protein 42
Gene Symbols ARHGAP42
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARHGAP42(Accession A6NI28), corresponding to amino acid residues around phosphorylated Tyr792.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143872, 500943, 71544
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.