Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-AS160 (Ser570) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105405 Supplier Invitrogen™ Supplier No. PA5105405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of AS160 only when phosphorylated at Ser570.

AS160 (TBC1D4) is a GTPase-activiting protein for RAB2A, RAB8A, RAB10, and RAB14. The isoform 2 version of this protein promotes the insulin-induced glucose transporter SLC2A4/GLUT4 translocation at the plasma membrane. This movement increases glucose uptake. Mutations associated with the gene cause Diabetes mellitus, non-insulin-dependent, 5 (NIDDM5).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-AS160 (Ser570)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TBC1D4
Gene Accession No. O60343, Q8BYJ6
Gene Alias 5930406J04Rik; A930035N22; Akt substrate of 160 kDa; AS160; AV295684; DKFZp779C0666; Kiaa0603; NIDDM5; OTTHUMP00000040818; RGD1561609; RP11-159J2.1; RP11-159J2.3; similar to TBC1 domain family member 4; TBC (Tre-2, BUB2, CDC16) domain-containing protein; TBC Tre-2; TBC1 domain family member 4; TBC1 domain family, member 4; TBC1D4
Gene Symbols TBC1D4
Host Species Rabbit
Immunogen A synthesized peptide derived from human TBC1D4(Accession O60343), corresponding to amino acid residues around phosphorylated Ser570.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 210789, 306117, 9882
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.