Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Ask1 (Ser83) Recombinant Rabbit Monoclonal Antibody (Ask1S83-G4), PE, Invitrogen™

Catalog No. PIMA537261
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537261 10 Tests
PIMA536974 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537261 Supplier Invitrogen™ Supplier No. MA537261
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

MAP3K5 (ASK1) is a serine/threonine kinase that is abundantly expressed in human heart and pancreas and are activated in response to various extracellular stimuli, including cytokines, growth factors and environmental stresses. A novel MAP kinase kinase kinase (MAPKKK) was identified and designated ASK1 (for apoptosis signal-regulating kinase 1) and MAPKKK5. ASK1 activates two different subgroups of MAPKK, MKK4 and MKK6, which in turn activates c-Jun N-terminal kinase (JNK) and p38 MAP kinase, respectively. ASK1/MAPKKK5 is activated by TNFR and Fas through the interaction with members of the TRAF family and Fas-associated protein Daxx. Overexpression of ASK1 induced apoptotic cell death and a catalytically inactive form of ASK1 inhibited TNF-a-induced apoptosis. ASK1 is expressed in variety of human and mouse tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Ask1 (Ser83)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone Ask1S83-G4
Conjugate PE
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene MAP3K5
Gene Accession No. O35099, Q99683
Gene Alias 7420452D20Rik; apoptosis signal regulating kinase 1; apoptosis signal-regulating kinase 1; ASK; ASK 1; ASK1; ASK-1; EC 2.7.11.25; kinase ASK1; M3K5; MAP/ERK kinase kinase 5; Map3k5; MAPK/ERK kinase kinase 5; MAPKKK5; MEK kinase 5; MEKK 5; Mekk5; mitogen activated protein kinase kinase kinase 5; mitogen-activated protein kinase kinase kinase 5; RGD1306565; RP3-325F22.4
Gene Symbols MAP3K5
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Ser83 of human phospho Ask1.
Purification Method Protein A/G
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26408, 365057, 4217
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.