Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ phospho-ATM (Ser1981), Mouse, Alexa Fluor 488, Clone: 10H11.E12,

Catalog No. 05740AF488 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 488
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
05-740-AF488 100 μL Alexa Fluor 488
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-740-AF488 Supplier MilliporeSigma™ Supplier No. 05740AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Serine-protein kinase ATM (EC 2.7.11.1; UniProt Q13315; also known as A-T mutated, Ataxia telangiectasia mutated) is encoded by the ATM (also known as AT) gene (Gene ID 472) in human. Ataxia telangiectasia mutated kinase (ATM) and ataxia telangiectasia and Rad3-related kinase (ATR) are related kinases that regulate cell cycle checkpoints and DNA repair. Mutation in the ATM gene results in the autosomal recessive disease ataxia telangiectasia (AT). Known ATM substrates include p53, p95/NBS1, MDM2, Chk2, BRCA1, CtIP, 4E-BP1, and Chk1. The S/TQ sequence constitutes the essential substrates phosphorylation site motif. Hydrophobic amino acids at positions -3 and -1, and negatively charged amino acids at position +1 are positive determinants for substrate phosphorylation, while positively charged residues surrounding the S/TQ are negative determinants. The complex phenotypes of cells derived from patients with AT suggests that ATM has additional cellular substrates. ATM is present as an inactive homodimer or multimer prior to activation. DNA double-stranded breaks induced by ionizing radiation (IR) cause rapid ATM autophosphorylation at Ser1981 in human or the Ser1987 equivalent in mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-ATM (Ser1981)
Applications Immunocytochemistry
Classification Monoclonal
Clone 10H11.E12
Concentration Please refer to lot specific datasheet.
Conjugate Alexa Fluor 488
Formulation Purified mouse monoclonal IgG1κ antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % Sodium Azide.
Gene ATM, AT
Gene Accession No. Q13315
Host Species Mouse
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 1974-1988 (SLAFEEG[pS]QSTTISS) of human ATM. The immunizing sequence is 11/12 identical to the corresponding mouse and rat sequence.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_000042
Test Specificity Reacts with ATM phosphorylated at serine 1981. Clone 10H11.E12 is covered by US patent No. 6,916,627 and 7,108,992.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.