Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ phospho-ATM (Ser1981), Mouse, Unlabeled, Clone: 10H11.E12,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MAB3806C
Description
Serine-protein kinase ATM (EC 2.7.11.1; UniProt Q13315; also known as A-T mutated, Ataxia telangiectasia mutated) is encoded by the ATM (also known as AT) gene (Gene ID 472) in human. Ataxia telangiectasia mutated kinase (ATM) and ataxia telangiectasia and Rad3-related kinase (ATR) are related kinases that regulate cell cycle checkpoints and DNA repair. Mutation in the ATM gene results in the autosomal recessive disease ataxia telangiectasia (AT). Known ATM substrates include p53, p95/NBS1, MDM2, Chk2, BRCA1, CtIP, 4E-BP1, and Chk1. The S/TQ sequence constitutes the essential substrates phosphorylation site motif. Hydrophobic amino acids at positions -3 and -1, and negatively charged amino acids at position +1 are positive determinants for substrate phosphorylation, while positively charged residues surrounding the S/TQ are negative determinants. The complex phenotypes of cells derived from patients with AT suggests that ATM has additional cellular substrates. ATM is present as an inactive homodimer or multimer prior to activation. DNA double-stranded breaks induced by ionizing radiation (IR) cause rapid ATM autophosphorylation at Ser1981 in human or the Ser1987 equivalent in mouse.
Specifications
| phospho-ATM (Ser1981) | |
| Monoclonal | |
| Please refer to lot specific datasheet. | |
| Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide. | |
| Q13315 | |
| Synthetic peptide corresponding to a.a. 1974-1988 of human ATM with phosphorylated Ser1981 (SLAFEEG[pS]QSTTISS). | |
| 100 μg | |
| Epigenetics & Nuclear Function | |
| NP_000042 | |
| Human | |
| Purified |
| Immunocytochemistry, Immunoprecipitation, Western Blot | |
| 10H11.E12 | |
| Unconjugated | |
| ATM, AT | |
| Mouse | |
| Protein G Purified | |
| RUO | |
| Primary | |
| Reacts with ATM Kinase phosphorylated at serine 1981. Clone 10H11.E12 is covered by US patent No. 6,916,627 and 7,108,992. | |
| Stable for 1 year at 2°-8°C from date of receipt. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction