Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-ATR (Thr1989) Recombinant Rabbit Monoclonal Antibody (PS01-18)

Catalog No. FEMA550716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550716 Supplier Invitrogen™ Supplier No. MA550716

Rabbit Recombinant Monoclonal Antibody

Positive control: HeLa cell lysate, HeLa treated with Hydroxyurea cell lysate, HeLa treated with 4mM Hydroxyurea for 20 hours. Predicted band size: 301 kDa Subcellular Location: Chromosome, Nucleus.

The protein encoded by this gene is a serine/threonine kinase and DNA damage sensor, activating cell cycle checkpoint signaling upon DNA stress. The encoded protein can phosphorylate and activate several proteins involved in the inhibition of DNA replication and mitosis, and can promote DNA repair, recombination, and apoptosis. This protein is also important for fragile site stability and centrosome duplication. Defects in this gene are a cause of Seckel syndrome 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-ATR (Thr1989)
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone PS01-18
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ATR
Gene Accession No. Q13535
Gene Alias 2310008J16Rik; 2810405N18Rik; ataxia telangiectasia and Rad3 related; ataxia telangiectasia and Rad3-related protein; ATR; ATR serine/threonine kinase; EC 2.7.11.1; FCTCS; FRAP-related protein 1; FRAP-related protein-1; FRP1; I79_000761; kinase ATR; MEC1; MEC1, mitosis entry checkpoint 1, homolog; protein kinase ATR; SCKL; SCKL1; Serine/threonine-protein kinase ATR; TEM8
Gene Symbols ATR
Host Species Rabbit
Immunogen Synthetic peptide surrounding Thr1989 of human ATR/2644.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 545
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.