Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Aurora A/B/C (Thr288, Thr232, Thr198) Recombinant Rabbit Monoclonal Antibody (AuroraABC-CC12), Invitrogen™

Catalog No. PIMA537398
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537398 20 μL
PIMA537152 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537398 Supplier Invitrogen™ Supplier No. MA537398
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Serine/threonine-protein kinase component of the chromosomal passenger complex (CPC), a complex that acts as a key regulator of mitosis. The CPC complex has essential functions at the centromere in ensuring correct chromosome alignment and segregation and is required for chromatin-induced microtubule stabilization and spindle assembly. Involved in the bipolar attachment of spindle microtubules to kinetochores and is a key regulator for the onset of cytokinesis during mitosis. Required for central/midzone spindle assembly and cleavage furrow formation. Key component of the cytokinesis checkpoint, a process required to delay abscission to prevent both premature resolution of intercellular chromosome bridges and accumulation of DNA damage: phosphorylates CHMP4C, leading to retain abscission-competent VPS4 (VPS4A and/or VPS4B) at the midbody ring until abscission checkpoint signaling is terminated at late cytokinesis (PubMed:22422861, PubMed:24814515). AURKB phosphorylates the CPC complex subunits BIRC5/survivin, CDCA8/borealin and INCENP. Phosphorylation of INCENP leads to increased AURKB activity. Other known AURKB substrates involved in centromeric functions and mitosis are CENPA, DES/desmin, GPAF, KIF2C, NSUN2, RACGAP1, SEPT1, VIM/vimentin, GSG2/Haspin, and histone H3. A positive feedback loop involving GSG2 and AURKB contributes to localization of CPC to centromeres. Phosphorylation of VIM controls vimentin filament segregation in cytokinetic process, whereas histone H3 is phosphorylated at 'Ser-10' and 'Ser-28' during mitosis (H3S10ph and H3S28ph, respectively). A positive feedback between GSG2 and AURKB contributes to CPC localization. AURKB is also required for kinetochore localization of BUB1 and SGOL1. Phosphorylation of p53/TP53 negatively regulates its transcriptional activity. Key regulator of active promoters in resting B- and T-lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Aurora A/B/C (Thr288, Thr232, Thr198)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone AuroraABC-CC12
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.10% BSA, 50% glycerol and 0.02% sodium azide
Gene Aurka
Gene Accession No. O14965, Q96GD4, Q9UQB9
Gene Alias Aik; Aik2; AIM1; AIM-1; Airk; AIRK1; AIRK2; AL022959; ARK1; ARK-1; ARK2; ARK-2; AU019385; AURA; AurB; AURKA; AURKB; aurkb-sv1; aurkb-sv2; Aurora 1; aurora 2; aurora A; aurora- and Ipl1-like midbody-associated protein 1; aurora B; aurora family kinase 1; Aurora kinase A; aurora kinase B; aurora kinase B-Sv1; aurora kinase B-Sv2; aurora/IPL1-like kinase; aurora/IPL1-related kinase 1; Aurora/IPL1-related kinase 2; aurora-1; aurora-A; aurora-B; aurora-related kinase 1; aurora-related kinase 2; AW539821; Ayk1; Breast tumor-amplified kinase; Btak; IAK; IAK1; IPL1; ipl1- and aurora-related kinase 1; PPP1R47; PPP1R48; protein phosphatase 1, regulatory subunit 47; protein phosphatase 1, regulatory subunit 48; ratAurA; serine/threonine kinase 12; serine/threonine kinase 5; serine/threonine kinase 6; serine/threonine protein kinase 15; serine/threonine protein kinase 6; serine/threonine-protein kinase 12; Serine/threonine-protein kinase 15; Serine/threonine-protein kinase 5; serine/threonine-protein kinase 6; Serine/threonine-protein kinase aurora-A; serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase Ayk1; STK1; STK-1; Stk12; Stk15; Stk5; STK6; STK7
Gene Symbols Aurka, Aurkb, AURKC
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding human Aurora A (Thr288)/Aurora B (Thr232)/Aurora C (Thr198).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6790, 6795, 9212
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.