Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Aurora B (Thr232) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538557
Description
Aurora kinases belong to a highly conserved family of mitotic serine/threonine kinases with three members identified among mammals: Aurora A, B, and C. Studies on the temporal expression pattern and subcellular localization of Aurora kinases in mitotic cells suggest an association with mitotic structure. Aurora kinase functional influences span from G2 phase to cytokinesis and may be involved in key cell cycle events such as centrosome duplication, chromosome bi-orientation and segregation, cleavage furrow positioning, and ingression.
Specifications
Phospho-Aurora B (Thr232) | |
Polyclonal | |
Unconjugated | |
AURKB | |
Aurora kinase B, Aurora 1, Aurora- and IPL1-like midbody-associated protein 1, AIM-1, Aurora/IPL1-related kinase 2, ARK-2, Aurora-related kinase 2, STK-1, Serine/threonine-protein kinase 12, Serine/threonine-protein kinase 5,Serine/threonine-protein kinase aurora-B, AURKB, AIK2, AIM1, AIRK2, ARK2, STK1, STK12, STK5, Aurora kinase C, Aurora 3, Aurora/IPL1-related kinase 3, ARK-3, Aurora-related kinase 3, Aurora/IPL1/Eg2 protein 2, Serine/threonine-protein kinase 13, Serine/threonine-protein kinase aurora-C, AURKC, AIE2, AIK3, AIRK3, ARK3, STK13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114592, 20877, 9212 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96GD4, O70126, O55099 | |
AURKB | |
A synthetic phosphopeptide derived from human AurB around the phosphorylation site of Thr232 (R-K-TP-M-C) | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction