Learn More
Phospho-AXL/UFO (Tyr770, Tyr812, Tyr857) Polyclonal Antibody, Abbomax
Rabbit Polyclonal Antibody
Supplier: Abbomax Inc 620970

Description
Positive control: Hela cells Cellular location: Membrane.
Phospho-AXL/UFO (Tyr770, Tyr812, Tyr857) Polyclonal antibody specifically detects Phospho-AXL/UFO (Tyr770, Tyr812, Tyr857) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
Phospho-AXL/UFO (Tyr770, Tyr812, Tyr857) | |
Polyclonal | |
Unconjugated | |
AXL | |
Adhesion-related kinase; AI323647; ARK; AXL; AXL oncogene; AXL receptor tyrosine kinase; AXL transforming sequence/gene; AZF1; JTK11; oncogene AXL; sAXL; soluble AXL; Tyro7; Tyrosine-protein kinase receptor UFO; Ufo; ufo oncogene homolog | |
Multiple phosphorylated sites of a panel synthetic peptides surrounding to the epitope -LDGLY*ALMSR-; EPDEILY*VNMDE-; EVHPAGRY*VL- with a phosphorylation site at Y770, Y812, Y857 of human AXL protein | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
P30530, Q00993 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26362, 308444, 558 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.