Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Axl (Y779) PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems IC6965P025
This item is not returnable.
View return policy
Description
Axl Monoclonal specifically detects Axl in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Axl (p Tyr779) | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
AXL | |
Phosphopeptide containing the human Axl Y779 site | |
25 Tests | |
Primary | |
Detects human Phospho-Axl (Y779) in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
713610 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
AI323647, ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
558 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction