Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-B-Raf (Thr401) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986418
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6418 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6418 Supplier Bioss Supplier No. BS5224R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

B-raf belongs to the raf/mil family of serine/threonine protein kinases. This protein plays a role in regulating the MAP kinase/ERKs signaling pathway, which affects cell division, differentiation, and secretion. Mutations in the gene are associated with cardiofaciocutaneous syndrome, a disease characterized by heart defects, mental retardation and a distinctive facial appearance. Mutations in this gene have also been associated with various cancers, including non-Hodgkin lymphoma, colorectal cancer, malignant melanoma, thyroid carcinoma, non-small cell lung carcinoma, and adenocarcinoma of lung. A pseudogene, which is located on chromosome X, has been identified for this gene.

Specifications

Antigen Phospho-B-Raf (Thr401)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BRAF
Gene Accession No. P15056
Gene Alias 94 kDa B-raf protein; 9930012E13Rik; AA120551; AA387315; AA473386; BRAF; B-raf; B-raf protein; B-Raf proto-oncogene serine/threonine-protein kinase; B-Raf proto-oncogene serine/threonine-protein kinase (p94); B-Raf proto-oncogene, serine/threonine kinase; Braf transforming gene; BRAF1; B-RAF1; Braf2; Braf-2; C230098H17; C87398; C-RMIL; D6Ertd631e; FLJ95109; MGC126806; murine sarcoma viral (v-raf) oncogene homolog B1; NS7; p94; Proto-oncogene B-Raf; Proto-oncogene c-Rmil; RAFB1; RMIL; rmil serine/threonine-protein kinase; serine/threonine kinase; Serine/threonine-protein kinase B-raf; Serine/threonine-protein kinase Rmil; v-raf murine sarcoma viral oncogene homolog B; v-Raf murine sarcoma viral oncogene homolog B1
Gene Symbols BRAF
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human B-Raf around the phosphorylation site of Thr401.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 673
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.