Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-B-Raf (Thr598) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA512836
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA512836 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA512836 Supplier Invitrogen™ Supplier No. PA512836
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine, chicken, mouse and rat based on sequence homology.

BRAF is involved in the transduction of mitogenic signals from the cell membrane to the nucleus. It is activated by somatic point mutation in human cancer, and is thought to play a role in the postsynaptic responses of hippocampal neuron. This protein plays a role in regulating the MAP kinase/ERKs signaling pathway, which affects cell division, differentiation, and secretion. Mutations in B-Raf have also been associated with various cancers, including non-Hodgkin lymphoma, colorectal cancer, malignant melanoma, thyroid carcinoma, non-small cell lung carcinoma, and adenocarcinoma of lung. A pseudogene, which is located on chromosome X, has been identified for B-Raf.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-B-Raf (Thr598)
Applications Dot Blot, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene BRAF
Gene Accession No. P15056
Gene Alias 94 kDa B-raf protein; 9930012E13Rik; AA120551; AA387315; AA473386; Braf; B-raf; B-raf protein; B-raf protein isoform 1; B-raf protein isoform 2; B-Raf proto-oncogene serine/threonine-protein kinase; B-Raf proto-oncogene serine/threonine-protein kinase (p94); B-Raf proto-oncogene, serine/threonine kinase; Braf transforming gene; BRAF1; B-RAF1; Braf2; Braf-2; C230098H17; C87398; C-RMIL; D6Ertd631e; FLJ95109; MGC126806; murine sarcoma viral (v-raf) oncogene homolog B1; NS7; p94; Proto-oncogene B-Raf; proto-oncogene c-Rmil; RAFB1; RMIL; rmil serine/threonine-protein kinase; serine/threonine kinase; serine/threonine-protein kinase B-raf; Serine/threonine-protein kinase Rmil; v-raf murine sarcoma viral oncogene homolog B; v-raf murine sarcoma viral oncogene homolog B1
Gene Symbols BRAF
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding T598 of human BRAF.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 673
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.