Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-BAD (Ser112) Recombinant Rabbit Monoclonal Antibody (BADS112-B9)
GREENER_CHOICE

Catalog No. PIMA537058
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537058 20 μL
PIMA528043 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537058 Supplier Invitrogen™ Supplier No. MA537058
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

The protein encoded by BAD is a member of the BCL-2 family. BCL-2 family members are known to be regulators of programmed cell death. This protein positively regulates cell apoptosis by forming heterodimers with BCL-xL and BCL-2, and reversing their death repressor activity. Proapoptotic activity of this protein is regulated through its phosphorylation. Protein kinases AKT and MAP kinase, as well as protein phosphatase calcineurin were found to be involved in the regulation of this protein. Alternative splicing of this gene results in two transcript variants which encode the same isoform.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-BAD (Ser112)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone BADS112-B9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.1% BSA and 0.02% sodium azide; pH 7.4
Gene BAD
Gene Accession No. O35147, Q61337, Q92934
Gene Alias AI325008; BAD; Bbc2; Bbc6; Bcl2 antagonist of cell death; BCL2 associated agonist of cell death; bcl-2 associated death agonist; Bcl2-antagonist of cell death; BCL2-antagonist of cell death protein; bcl2-associated agonist of cell death; bcl2-associated death promoter; bcl-2-binding component 6; BCL2-binding component 6; BCL2-binding protein; BCL2L8; bcl2-L-8; Bcl-2-like protein 8; BCL-X/BCL-2 binding protein; bcl-XL/Bcl-2-associated death promoter; hypothetical protein LOC615013; OTTMUSP00000017561; OTTMUSP00000022400
Gene Symbols BAD
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Ser112 of human phospho BAD.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12015, 572, 64639
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.