Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-BAD (Ser112) Recombinant Rabbit Monoclonal Antibody (BADS112-B9), APC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
$245.00 - $598.50
Specifications
Antigen | Phospho-BAD (Ser112) |
---|---|
Clone | BADS112-B9 |
Applications | Flow Cytometry |
Classification | Recombinant Monoclonal |
Conjugate | APC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA537320
|
Invitrogen™
MA537320 |
10 Tests |
Each for $245.00
|
|
|||||
PIMA537060
|
Invitrogen™
MA537060 |
100 Tests |
Each for $598.50
|
|
|||||
Description
The protein encoded by BAD is a member of the BCL-2 family. BCL-2 family members are known to be regulators of programmed cell death. This protein positively regulates cell apoptosis by forming heterodimers with BCL-xL and BCL-2, and reversing their death repressor activity. Proapoptotic activity of this protein is regulated through its phosphorylation. Protein kinases AKT and MAP kinase, as well as protein phosphatase calcineurin were found to be involved in the regulation of this protein. Alternative splicing of this gene results in two transcript variants which encode the same isoform.Specifications
Phospho-BAD (Ser112) | |
Flow Cytometry | |
APC | |
Rabbit | |
Human, Mouse, Rat | |
O35147, Q61337, Q92934 | |
12015, 572, 64639 | |
A synthetic phospho-peptide corresponding to residues surrounding Ser112 of human phospho BAD. | |
Primary | |
4°C | |
BAD |
BADS112-B9 | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with 0.20% BSA and 0.09% sodium azide | |
AI325008; BAD; Bbc2; Bbc6; Bcl2 antagonist of cell death; BCL2 associated agonist of cell death; bcl-2 associated death agonist; Bcl2-antagonist of cell death; BCL2-antagonist of cell death protein; bcl2-associated agonist of cell death; bcl2-associated death promoter; bcl-2-binding component 6; BCL2-binding component 6; BCL2-binding protein; BCL2L8; bcl2-L-8; Bcl-2-like protein 8; BCL-X/BCL-2 binding protein; bcl-XL/Bcl-2-associated death promoter; hypothetical protein LOC615013; OTTMUSP00000017561; OTTMUSP00000022400 | |
BAD | |
IgG λ | |
Protein A/G | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title