Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Band3 (Tyr21) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982350
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2350 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2350 Supplier Bioss Supplier No. BS12569R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The Band 3 protein is the most abundant integral protein of human erythrocyte membranes and functions as the major anion transporter polypeptide of erythrocytes. Band 3 is a 90-100 kDa protein, its microheterogeneity is ascribed mainly to the structural heterogeneity of the carbohydrate moiety. The C-terminal portion of the molecule spans the membrane several times, and is responsible for anion transport. The N-terminal portion, comprising almost half of the polypeptide chain of Band 3, is located inside the red cell and interacts with cytoskeletal and cytoplasmic proteins such as ankyrin.

Specifications

Antigen Phospho-Band3 (Tyr21)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLC4A1
Gene Accession No. P02730
Gene Alias AE 1; Ae1; Anion exchange protein 1; anion exchanger 1; anion exchanger-1; Band 3 anion transport protein; BND3; C13orf8; CAMP; CD233; CHAMP; CHC; DI; Diego blood group; EMPB3; EPB3; erythrocyte membrane protein band 3; erythroid anion exchange protein; FR; Froese blood group; l11Jus51; MEB3; MGC116750; MGC116753; MGC126619; MGC126623; RTA1A; SAO; Slc4a1; solute carrier family 4 (anion exchanger), member 1; solute carrier family 4 (anion exchanger), member 1 (Diego blood group); Solute carrier family 4 member 1; solute carrier family 4 member 1 (Diego blood group); solute carrier family 4, anion exchanger, member 1 (erythrocyte membrane protein band 3); solute carrier family 4, anion exchanger, member 1 (erythrocyte membrane protein band 3, Diego blood group); solute carrier family 4, anion exchanger, number 1; Solute carrier family 4, member 1, anion exchange protein 1 (kidney band 3); SPH4; SW; Swann blood group; Waldner blood group; WD; WD1; WR; Wright blood group; ZNF828
Gene Symbols SLC4A1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human Band3 around the phosphorylation site of Tyr21.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6521
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.