Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Bax (Ser184) Polyclonal Antibody, FITC, Bioss
SDP

Catalog No. 502279519
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-227-9519 Supplier Bioss Supplier No. BS3010RFITC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-Bax (Ser184) Polyclonal antibody specifically detects Phospho-Bax (Ser184) in Human, Mouse, Rat samples. It is validated for Flow Cytometry

Specifications

Antigen Phospho-Bax (Ser184)
Applications Flow Cytometry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate FITC
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene BAX
Gene Accession No. Q07812, Q07813, Q63690
Gene Alias apoptosis regulator BAX; Apoptosis regulator Bcl-X; Bax; Bax zeta; BAX-ALPHA; Bax-alpha protein; Baxdelta2G9; Baxdelta2G9omega; Baxdelta2omega; BCL2 associated X protein; BCL2 associated X, apoptosis regulator; BCL2 like 1; BCL2-associated protein; Bcl-2-associated protein Bax; Bcl2-associated protein Bax; Bcl-2-associated X protein; BCL2-associated X protein; BCL2-associated X protein omega; BCL2L; BCL2L1; Bcl2-L-1; BCL2L4; Bcl2-L-4; BCL2-like 1; bcl-2-like protein 1; Bcl-2-like protein 4; BCLX; Bcl-X; BCL-XL/S; PPP1R52; protein phosphatase 1, regulatory subunit 52
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human Bax around the phosphorylation site of Ser184
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12028, 24887, 581
Target Species Human, Mouse, Rat
Content And Storage -20° C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.