Learn More
Invitrogen™ Phospho-BCLAF1 (Ser531) Polyclonal Antibody

Description
Antibody detects endogenous levels of BCLAF1 only when phosphorylated at Ser531.
Specifications
Specifications
| Antigen | Phospho-BCLAF1 (Ser531) |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | BCLAF1 |
| Gene Accession No. | Q8K019, Q9NYF8 |
| Gene Alias | 2610102K23Rik; 2700025J07Rik; 2810454G14Rik; 5730534O06Rik; Aa2-041; AI450190; AW556225; BCL2 associated transcription factor 1; bcl-2-associated transcription factor 1; BCL2-associated transcription factor 1; Bcl-2-associated transcription factor 1-like protein; BCLAF1; BCLAF1 and THRAP3 family member 1; bK211L9.1; BP-1; BTF; Btf-s; KIAA0164; mKIAA0164 |
| Gene Symbols | BCLAF1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.