Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Bim (Thr56, Thr116) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564655
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564655 Supplier Invitrogen™ Supplier No. PA564655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-Bim (Thr56, 116) Polyclonal Antibody detects endogenous levels of Bim only when phosphorylated at Thr56, 116.

BIM belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. The protein encoded by this gene contains a Bcl-2 homology domain 3 (BH3). It has been shown to interact with other members of the BCL-2 protein family, including BCL2, BCL2L1/BCL-X(L), and MCL1, and to act as an apoptotic activator. The expression of this gene can be induced by nerve growth factor (NGF), as well as by the forkhead transcription factor FKHR-L1, which suggests a role of this gene in neuronal and lymphocyte apoptosis. Transgenic studies of the mouse counterpart suggested that this gene functions as an essential initiator of apoptosis in thymocyte-negative selection. Several alternatively spliced transcript variants of this gene have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Bim (Thr56, Thr116)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BCL2L11
Gene Accession No. O43521, O54918, O88498
Gene Alias 1500006F24Rik; BAM; Bcl2 interacting mediator of cell death; bcl-2 interacting mediator of cell death; bcl-2 interacting protein Bim; BCL2 like 11; Bcl-2 like 11; Bcl-2 related apoptotic gene product BimL; bcl2-interacting mediator of cell death; BCL2L11; Bcl2-L-11; BCL2-like 11 (apoptosis facilitator); bcl-2-like protein 11; bcl-2-like protein 11-like; bcl-2-related ovarian death agonist; bcl-2-related ovarian death protein; BIM; Bim EL; Bim-A; BIM-alpha6; BIM-beta6; BIM-beta7; BIMEL; BimEL protein; BimL; BOD; OTTHUMP00000197013; OTTHUMP00000204247; Unknown (protein for MGC:134444)
Gene Symbols BCL2L11
Host Species Rabbit
Immunogen A synthesized peptide derived from human Bim (Phospho-Thr56/116).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10018, 12125, 64547
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.