Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-BLM (Ser1290)Rabbit anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501992243
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2243 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-2243 Supplier Rockland Immunochemicals Supplier No. 600401J55
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-BLM pS1290 is directed against the phosphorylated form of human S1290 residue. A BLAST analysis was used to suggest cross-reactivity with human and mouse based on 100% sequence homology. Cross-reactivity with BLM pS1290 from other sources has not been determined.

The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.

Specifications

Antigen Phospho-BLM (Ser1290)
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and no preservative; pH 7.2
Gene BLM
Gene Accession No. P54132
Gene Alias BLM; Bloom syndrome homolog; Bloom syndrome protein; Bloom syndrome protein homolog; Bloom syndrome RecQ like helicase; Bloom syndrome, RecQ helicase-like; BS; DNA helicase, RecQ-like type 2; mBLM; MGC126616; MGC131618; MGC131620; RecQ helicase homolog; RecQ protein-like 3; RECQ2; RECQL2; RECQL3
Gene Symbols BLM
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 641
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.