Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-BLNK (Tyr84) Recombinant Rabbit Monoclonal Antibody (BLNKY84-H4), APC, Invitrogen™
GREENER_CHOICE

Catalog No. PIMA537087
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537087 100 Tests
PIMA537337 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537087 Supplier Invitrogen™ Supplier No. MA537087
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

B cell linker protein (Blnk) is a protein encoded by the Blnk gene located on chromosome 10 in humans. It is also known as SLP 65 (Src homology 2 domain-containing leukocyte protein of 65 kDa). It derives its name from being an adaptor protein in the B cell receptor signaling cascade with several supporting studies showing improper development of B cells in the absence of Blnk. Blnk undergoes tyrosine phosphorylation by Syk, a B cell receptor activated kinase, which provides a platform for interaction of signaling intermediaries such as the Vav GEF and Grb2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-BLNK (Tyr84)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone BLNKY84-H4
Conjugate APC
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene BLNK
Gene Accession No. Q8WV28
Gene Alias AGM4; B cell adaptor containing SH2 domain; B cell linker; B cell linker protein; BASH; bca; B-cell activation; B-cell adapter containing a SH2 domain protein; B-cell adapter containing a Src homology 2 domain protein; B-cell linker; B-cell linker protein; Blnk; BLNK-S; Cytoplasmic adapter protein; LY57; Ly-57; lymphocyte antigen 57; Lyw-57; MGC111051; SLP65; Slp-65; Src homology [SH2] domain-containing leukocyte protein of 65 kD; Src homology 2 domain-containing leukocyte protein of 65 kDa
Gene Symbols BLNK
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Tyr84 of human phospho BLNK.
Purification Method Protein A/G
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29760
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.