Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-BMAL1 (Ser49) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105374
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105374 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105374 Supplier Invitrogen™ Supplier No. PA5105374
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of BMAL1 only when phosphorylated at Ser49.

The protein encoded by this gene is a basic helix-loop-helix protein that forms a heterodimer with CLOCK. This complex binds an E-box upstream of the PER1 gene, activating this gene and possibly other circadian rhythym-associated genes. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-BMAL1 (Ser49)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Arntl
Gene Accession No. O00327, Q9EPW1, Q9WTL8
Gene Alias Arnt3; ARNTL; ARNT-like protein 1, brain and muscle; aryl hydrocarbon receptor nuclear translocator like; aryl hydrocarbon receptor nuclear translocator-like; aryl hydrocarbon receptor nuclear translocator-like protein 1; basic-helix-loop-helix-PAS orphan MOP3; basic-helix-loop-helix-PAS protein MOP3; bHLHe5; bHLH-PAS protein JAP3; BMAL1; Bmal1 protein; BMAL1b; bmal1b'; BMAL1c; Brain and muscle ARNT-like 1; brain and muscle Arnt-like protein 1; CG8727 PA; class E basic helix-loop-helix protein 5; cycle; JAP3; LOW QUALITY PROTEIN: aryl hydrocarbon receptor nuclear translocator-like protein 1; member of PAS protein 3; member of PAS superfamily 3; MGC47515; MOP3; PAS domain-containing protein 3; PASD3; testis tissue sperm-binding protein Li 50e; tic
Gene Symbols Arntl
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARNTL(Accession O00327), corresponding to amino acid residues around phosphorylated Ser49.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11865, 29657, 406
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.