Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-BRAF35 (Thr401) Recombinant Rabbit Monoclonal Antibody (5G6Q5)
GREENER_CHOICE

Catalog No. PIMA535877
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535877 Supplier Invitrogen™ Supplier No. MA535877
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: SATPP.

Required for correct progression through G2 phase of the cell cycle and entry into mitosis. Required for RCOR1/CoREST mediated repression of neuronal specific gene promoters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-BRAF35 (Thr401)
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 5G6Q5
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BRAF
Gene Accession No. P15056, Q9P0W2
Gene Alias AW610687; BRAF25; BRAF35; BRCA2-associated factor 35; fc85b02; FLJ26127; high mobility group 20 B; high mobility group 20B; high mobility group box domain containing; high mobility group protein 20 B; high-mobility group 20B; HMG box domain containing 2; HMG box-containing protein 20B; HMG domain-containing protein 2; HMG domain-containing protein HMGX2; HMG20B; Hmgx2; HMGXB2; hypothetical protein LOC553572; PP7706; pp8857; si:dkey-110c1.6; SMARCE1R; SMARCE1-related protein; SOXL; Sox-like transcriptional factor; Structural DNA-binding protein BRAF35; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; LOW QUALITY PROTEIN: SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E, member 1-related; Unknown (protein for MGC:134468); wu:fc85b02; zgc:110001
Gene Symbols BRAF, HMG20B
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around T401 of human-B Raf (NP_0043242).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10362, 114486, 362825, 673
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.