Learn More
phospho Brd4 (Ser492/Ser494) Rabbit anti-Human, Mouse, Rat, Porcine, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho Brd4 (Ser492/Ser494) |
| Applications | Immunocytochemistry, Dot Blot, Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Gene Symbols | BRD4,HUNK1 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to 14 amino acids surrounding Ser492/Ser494 from the internal N-terminal half of human phospho Brd4 protein. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.