Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-BRD4 (Thr204) Recombinant Rabbit Monoclonal Antibody (3V5R1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542961
This item is not returnable.
View return policy
Description
Positive test controls include: HeLa, C6, NIH/3T3. The target is usually found in the following locations: Chromosome, Nucleus. Immunogen sequence: ASTPP.
BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; the longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of the cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.
Specifications
Phospho-BRD4 (Thr204) | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
O60885 | |
BRD4 | |
A synthetic phosphorylated peptide around T204 of human BRD4 (NP_490597.1). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
3V5R1 | |
Unconjugated | |
BRD4 | |
Brd4; BRD4-NUT FUSION; Brd5; bromodomain containing 4; bromodomain-containing 4; bromodomain-containing 5; bromodomain-containing protein 4; bromodomain-containing protein 4; LOW QUALITY PROTEIN: bromodomain-containing protein 4; CAP; chromosome-associated protein; fsh-like protein; HUNK1; HUNKI; Mcap; mitotic chromosome-associated protein; Protein HUNK1; WI-11513 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23476, 362844 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction