Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-BRK (Tyr447) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538413
Description
The protein encoded by this gene is a cytoplasmic nonreceptor protein kinase which may function as an intracellular signal transducer in epithelial tissues. Overexpression of this gene in mammary epithelial cells leads to sensitization of the cells to epidermal growth factor and results in a partially transformed phenotype. Expression of this gene has been detected at low levels in some breast tumors but not in normal breast tissue. The encoded protein has been shown to undergo autophosphorylation.
Specifications
Phospho-BRK (Tyr447) | |
Polyclonal | |
Unconjugated | |
PTK6 | |
BRK, breast tumor kinase, protein-tyrosine kinase 6, protein-tyrosine kinase BRK, tyrosine-protein kinase BRK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20459, 5753 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q13882, Q64434 | |
PTK6 | |
A synthetic phosphopeptide derived from human Breast Tumor Kinase around the phosphorylation site of Tyr447 (T-S-YP-E-N) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction